missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium oxalate hydrate, 99%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1228930

 View more versions of this product

Catalog No. AAA1228930



Description

Description

Applications
Potassium oxalate hydrate is used as a chelating agent, a purifying agent in the pharmaceutical industry and a precipitating agent in rare-earth metal processing. It finds application as a grinding agent, as a rust remover in metal treatment and as a bleaching agent in the textile and wood industry. It is also used in waste water treatment and as a scale remover in automobile radiators. Furthermore, it is used as a reagent in analytical chemistry for the determination of calcium and other heavy metals as well as reducing agents in photography. It is also involved in the preparation of SBA-15 (nanostructured SiO2) supported vanadium oxide catalyst.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with halogens, ammonia, cyanides and heavy metals.
Specifications

Specifications

Potassium oxalate hydrate
>300°C
6 to 8.5
99%
MFCD00150033
14,7651
QCPTVXCMROGZOL-UHFFFAOYSA-L
dipotassium;oxalate;hydrate
2724193
99%
Odorless
250g
6487-48-5
C2H2K2O5
3752576
potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
C(=O)(C(=O)[O-])[O-].O.[K+].[K+]
184.23
166.22 (Anhydrous)

Chemical Identifiers

6487-48-5
184.23
QCPTVXCMROGZOL-UHFFFAOYSA-L
2724193
C(=O)(C(=O)[O-])[O-].O.[K+].[K+]
C2H2K2O5
MFCD00150033
potassium oxalate monohydrate, potassium oxalate hydrate, oxalic acid potassium salt, ethanedioic acid, dipotassium salt, monohydrate, dipotassium oxalate hydrate, dipotassium hydrate oxalate, acmc-1bcm6, ksc495a4h, dipotassium oxalate monohydrate, dipotassium ethanedioate hydrate
dipotassium;oxalate;hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280-P301+P312-P312-P363-P322-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Specific measures (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : RO2885000

TSCA : TSCA

storageNote1 : Hygroscopic

SDS Documents