Please login to your online account to display your discounted pricing

Potassium Oxalate, For Dissolved Oxygen, Certified, 2.0% (w/v) ±0.1%, LabChem™

$39.09 - $39.09

Chemical Identifiers

Chemical Name or Material Potassium Oxalate
CAS 7732-18-5
CAS Max % 98
CAS 6487-48-5
CAS Max % 2
Name Note For Dissolved Oxygen
Purity Grade Certified Grade
Linear Formula K2C2O4·H2O
Formula Weight 184.24
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC198401

 
lab chem
LC198401
Poly Bottle 500mL Each for $39.09
Description & Specifications

Specifications

Purity Grade Certified Grade
Assay 2.0% ± 0.1% (w/v)
Formula Weight 184.24
Chemical Name or Material Potassium Oxalate
Decomposition Information Carbon monoxide; Carbon dioxide
Packaging Poly Bottle
Specific Gravity 1g/mL
Traceability to NIST Traceable to NIST
Name Note For Dissolved Oxygen
Physical Form Liquid
Density 1g/mL
Linear Formula K2C2O4·H2O
Color Clear, colorless
CAS 7732-18-5
Quantity 500mL
Solubility Information Soluble in water
CAS Max % 98
Health Hazard 2 GHS H Statement
Solution is not hazardous.
CAS 6487-48-5
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Identification Passes Test
CAS Max % 2
Recommended Storage Room Temperature
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.

Room Temperature