missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium osmium(VI) oxide dihydrate, >99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1264701

 View more versions of this product

Catalog No. AA1264701



Specifications

Specifications

Potassium osmium(VI) oxide dihydrate
99+%
MFCD00149919
potassium osmate vi dihydrate, potassium dioxidodioxoosmium dihydrate, potassium osmate dihydrate, unii-5m2bwq2tyj, potassium osmate vi dihydrate, 5m2bwq2tyj, osmate oso42-, dipotassium, dihydrate, t-4, dipotassium tetrahydroxodioxoosmate, potassium osmate k2oso2 oh 4, potassium osmate k2oso4 , dihydrate
O.O.[O-][Os](=O)(=O)[O-].[K+].[K+]
368.453
368.43 (332.40 Anhydrous)
White
10022-66-9
H4K2O6Os
Purple powder
DGODWNOPHMXOTR-UHFFFAOYSA-N
dipotassium;dioxido(dioxo)osmium;dihydrate
53393272
Powder
0.25g

Chemical Identifiers

10022-66-9
368.453
DGODWNOPHMXOTR-UHFFFAOYSA-N
53393272
O.O.[O-][Os](=O)(=O)[O-].[K+].[K+]
H4K2O6Os
MFCD00149919
potassium osmate vi dihydrate, potassium dioxidodioxoosmium dihydrate, potassium osmate dihydrate, unii-5m2bwq2tyj, potassium osmate vi dihydrate, 5m2bwq2tyj, osmate oso42-, dipotassium, dihydrate, t-4, dipotassium tetrahydroxodioxoosmate, potassium osmate k2oso2 oh 4, potassium osmate k2oso4 , dihydrate
dipotassium;dioxido(dioxo)osmium;dihydrate
Safety and Handling

Safety and Handling

H301-H331-H315-H319-H335: Toxic if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 243-247-1

Documents