Please login to your online account to display your discounted pricing

Potassium osmate(VI) dihydrate, 51.0-52.0% Os, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Potassium osmate(VI) dihydrate
Name Note 51.0-52.0% Os
CAS 10022-66-9
Assay 51.0 to 52.0% (Os)
Molecular Formula K2O4Os·2H2O
Linear Formula K2OsO4·2H2O
Formula Weight 368.42
MDL Number MFCD00149919
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC197660025

  SDS
thermo fisher scientific
197660025
Glass bottle 2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC197665000

  SDS
thermo fisher scientific
197665000
Glass bottle 500mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 368.42
Quantity 2.5g
Color Violet
EINECS Number 243-247-1
TSCA TSCA
Assay 51.0 to 52.0% (Os)
Molecular Formula K2O4Os·2H2O
Chemical Name or Material Potassium osmate(VI) dihydrate
Name Note 51.0-52.0% Os
Linear Formula K2OsO4·2H2O
MDL Number MFCD00149919
CAS 10022-66-9
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
May intensify fire; oxidizer.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep/Store away from clothing/ combustible materials.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep/Store away from clothing/ combustible materials.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
May intensify fire; oxidizer.