Learn More
Potassium orthovanadate, 99.9% (metals basis), Thermo Scientific Chemicals
$419.19 - $1469.27
Chemical Identifiers
CAS | 14293-78-8 |
---|---|
Molecular Formula | K3O4V |
Molecular Weight (g/mol) | 232.232 |
MDL Number | MFCD00210642 |
InChI Key | FBRINGOENSVSPU-UHFFFAOYSA-N |
Synonym | Potassium vanadate (ortho) |
PubChem CID | 22023817 |
IUPAC Name | tripotassium;trioxido(oxo)vanadium |
SMILES | [O-][V](=O)([O-])[O-].[K+].[K+].[K+] |
Description
It is employed as in chemical fertilizer industries and used as corrosion inhibitor. It is also used for ammonia synthesis. Used as a catalyst in oil & gas industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14293-78-8 | |
232.232 | |
FBRINGOENSVSPU-UHFFFAOYSA-N | |
22023817 | |
[O-][V](=O)([O-])[O-].[K+].[K+].[K+] |
K3O4V | |
MFCD00210642 | |
Potassium vanadate (ortho) | |
tripotassium;trioxido(oxo)vanadium |
Specifications
14293-78-8 | |
25 g | |
MFCD00210642 | |
Potassium vanadate (ortho) | |
FBRINGOENSVSPU-UHFFFAOYSA-N | |
tripotassium;trioxido(oxo)vanadium | |
22023817 | |
99.9% | |
Potassium orthovanadate |
Granules | |
K3O4V | |
UN3285 | |
Soluble in water. | |
[O-][V](=O)([O-])[O-].[K+].[K+].[K+] | |
232.232 | |
232.24 | |
Odorless | |
(metals basis) |
Safety and Handling
GHS H Statement
H331-H315-H319-H335
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H332-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
EINECSNumber : 238-220-6
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only