Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium, Oil based standard solution, Specpure™, K 1000μg/g

Chemical Identifiers

Chemical Name or Material Potassium Oil based standard solution
Name Note K 1000μg/g
Purity Grade Specpure™
MDL Number MFCD00133776
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1421618

  SDS
alfa aesar™
1421618
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 50g
Purity Grade Specpure™
MDL Number MFCD00133776
Chemical Name or Material Potassium Oil based standard solution
Physical Form Liquid
Name Note K 1000μg/g