Please login to your online account to display your discounted pricing

Potassium Nitrite (Crystalline/Certified ACS), Fisher Chemical

$313.77 - $313.77

Chemical Identifiers

Chemical Name or Material Potassium Nitrite
CAS 7758-09-0
Purity Grade Certified ACS Grade
Physical Form Solid
Assay ≥96%
Assay Percent Range ≥96 %
Molecular Formula KNO2
Formula Weight 85.1
MDL Number MFCD00011408
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

P267-250

  SDS
fisher scientific company
P267250
Glass Bottle 250g Each for $313.77
Case of 6 Each for $1,882.62
Description & Specifications

Specifications

Assay ≥96%
Melting Point 387°C
Formula Weight 85.1
Purity Grade Certified ACS Grade
Quantity 250g
DOT Information DOT Class 5.1, : Oxidizer
Physical Form Solid
pH 7 to 10
Color Off-white
Density 1.9150g/cm³
UN Number 1488
Iron (Fe) 0.001% max.
Chloride 0.03% max.
Sodium (Na) 0.5% max.
Assay Percent Range ≥96 %
Molecular Formula KNO2
Magnesium (Mg) 0.002% max.
MDL Number MFCD00011408
Insoluble Matter 0.01% max.
Identification Pass Test
Heavy Metals (as Pb) 0.001% max.
Packaging Glass Bottle
Sulfate 0.01% max.
Calcium (Ca) 0.005% max.
Chemical Name or Material Potassium Nitrite
ChemAlert Storage Symbol Yellow
CAS 7758-09-0
Safety and Handling