missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium nitrite, ACS, 96.0% min, Alfa Aesar™

Manufacturer:  Alfa Aesar 1424536

 View more versions of this product

Catalog No. AA1424536



Specifications

Specifications

Potassium nitrite
7758-09-0
KNO2
potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320
N(=O)[O-].[K+]
85.103
85.11
1.915g/mL
For ACS analysis
96.0% min
MFCD00011408
BXNHTSHTPBPRFX-UHFFFAOYSA-M
potassium;nitrite
516910
Crystalline
500g

Chemical Identifiers

7758-09-0
85.103
BXNHTSHTPBPRFX-UHFFFAOYSA-M
516910
N(=O)[O-].[K+]
KNO2
MFCD00011408
potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320
potassium;nitrite
Safety and Handling

Safety and Handling

EINECSNumber : 231-832-4

SDS Documents