Please login to your online account to display your discounted pricing

Potassium nitrite, 97%, extra pure, ACROS Organics™


Quantity 5kg

Manufacturer: thermo fisher scientific  AC453190050

 View more versions of this product

Catalog No. AC453190050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 85.1
Quantity 5kg
Chemical Name or Material Potassium nitrite
Name Note Extra Pure
Assay Percent Range 97%
Molecular Formula KNO2
MDL Number MFCD00011408
CAS 7758-09-0

Specifications

Chemical Name or Material Potassium nitrite
Name Note Extra Pure
CAS 7758-09-0
Assay Percent Range 97%
Molecular Formula KNO2
Formula Weight 85.1
MDL Number MFCD00011408