Please login to your online account to display your discounted pricing

Potassium nitrite, 97%, for analysis, ACROS Organics™


Quantity 5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC222700050

 View more versions of this product

Catalog No. AC222700050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystals
Formula Weight 85.1
Quantity 5kg
Color White to slightly yellow
Purity Grade For analysis
Merck Index 15, 7770
Fieser 10,329
Linear Formula KNO2
Chlorine (Cl) 100ppm max.
MDL Number MFCD00011408
Chemical Name or Material Potassium nitrite
Assay Percent Range 97%
Molecular Formula KNO2
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Arsenic (As) 1ppm max.
Iron (Fe) 10ppm max.
Packaging Plastic bottle
Lead (Pb) 10ppm max.
Sulfate 200ppm max.
CAS 7758-09-0
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Toxic if swallowed.
Very toxic to aquatic life.
May intensify fire; oxidizer.

Specifications

Chemical Name or Material Potassium nitrite
CAS 7758-09-0
Purity Grade For analysis
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula KNO2
Linear Formula KNO2
Formula Weight 85.1
MDL Number MFCD00011408
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Toxic if swallowed.
Very toxic to aquatic life.
May intensify fire; oxidizer.

SDS & Safety Documents