Learn More
Potassium nitrite, 97%, for analysis, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 222700010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium nitrite | |
7758-09-0 | |
Crystals | |
97% | |
KNO2 | |
10,329 | |
potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320 | |
BXNHTSHTPBPRFX-UHFFFAOYSA-M | |
potassium;nitrite | |
516910 | |
97% | |
Plastic bottle |
387.0°C | |
White to Yellow | |
1 kg | |
KNO2 | |
MFCD00011408 | |
15, 7770 | |
Solubility in water: 3000g/L water (20°C). Other solubilities: 413 g/100mL water (100°C), slightly soluble in alcohol, soluble in hot alcohol, very soluble in liquid nh3 | |
[K+].[O-]N=O | |
85.10 | |
85.1 | |
Analytical |
Chemical Identifiers
7758-09-0 | |
85.10 | |
BXNHTSHTPBPRFX-UHFFFAOYSA-M | |
516910 |
KNO2 | |
MFCD00011408 | |
potassium nitrite, nitrous acid, potassium salt, caswell no. 698, ccris 3959, potassium nitrite 1:1, epa pesticide chemical code 076203, nitrous acid, potassium salt 1:1, kaliumnitrit, acmc-1bi01, dsstox_cid_22320 | |
[K+].[O-]N=O |
Safety and Handling
GHS H Statement:
Toxic if swallowed.
Very toxic to aquatic life.
May intensify fire; oxidizer.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid release to the environment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 231-832-4
RUO – Research Use Only