Please login to your online account to display your discounted pricing

Potassium Nitrate Fine Crystal, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Potassium Nitrate
CAS 7757-79-1
Physical Form Fine Crystals
Molecular Formula KNO3
Formula Weight 101.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK6715212

 
avantor performance
671506
Poly 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK6715500

 
avantor performance
671504
POLYSTORMOR 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Fine Crystals
Formula Weight 101.09
CAS 7757-79-1
Packaging Poly
Molecular Formula KNO3
Chemical Name or Material Potassium Nitrate
Quantity 2.5kg