Please login to your online account to display your discounted pricing

Potassium Nitrate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$251.36 - $892.22

Chemical Identifiers

Chemical Name or Material Potassium Nitrate
CAS 7757-79-1
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula KNO3
Formula Weight 101.09
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003785

 
avantor performance
319005
Poly Bottle 2.5kg CALL N/A N/A
 Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

02003786

 
avantor performance
319001
Wide Mouth Amber Glass Bottle 500g CALL N/A N/A
 Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade Baker Analyzed™ ACS Reagent
Physical Form Crystals
Formula Weight 101.09
CAS 7757-79-1
Chemical Name or Material Potassium Nitrate
Packaging Poly Bottle
Specific Gravity 1L = 2.10kg
Molecular Formula KNO3
Quantity 2.5kg