missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Nitrate, ACS Grade, ≥ 99.0%, LabChem™

$115.20 - $115.20

Chemical Identifiers

CAS 7757-79-1
CAS Max % 100
Linear Formula KNO3
Formula Weight 101.1
UN Number UN1486
Chemical Name or Material Potassium Nitrate
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
LC198181 Lab Chem
LC198181
500g Each for $115.20
Specifications Description & Specifications

Chemical Identifiers

7757-79-1
KNO3
UN1486
ACS Grade
100
101.1
Potassium Nitrate

Specifications

7757-79-1
Crystalline
101.1
2,100kg/m3
Poly Bottle
2100kg/m3
Nitrogen oxides; Oxygen
334°C
Potassium Nitrate
ACS Grade
100
KNO3
UN1486
Passes Test
Soluble in water
Room Temperature
White
500g
≥99.0%

Safety and Handling

GHS H Statement
May intensify fire; oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep/Store away from clothing, combustible materials.
Take any precaution to avoid mixing with combustibles.
Avoid breathing dust.
Use only outdoors or in a well-ventilated area.
Wear eye protection, protective clothing, protective gloves, face protection.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing and wash before reuse.
If skin irritation occurs: Get medical advice/attention.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Store locked up in a well-ventilated place.
Keep container tightly closed.
Dispose of contents/container to comply with local, state and federal regulations.

Warning

EINECSNumber : 231-818-8