missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium nitrate, ACS, 99.0% min, Alfa Aesar™

For Pyrotechnics, Glass Manufacture, Tempering Steel

Manufacturer:  Alfa Aesar 13443A3

 View more versions of this product

Catalog No. AA13443A3

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Potassium nitrate
7757-79-1
KNO3
White crystalline
FGIUAXJPYTZDNR-UHFFFAOYSA-N
potassium;nitrate
24434
101.11
ACS
White
For ACS analysis
99.0% min.
MFCD00011409
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
[N+](=O)([O-])[O-].[K+]
101.102
CHEBI:63043
Crystals
Odorless
2kg

Chemical Identifiers

7757-79-1
101.102
FGIUAXJPYTZDNR-UHFFFAOYSA-N
24434
potassium;nitrate
KNO3
MFCD00011409
potassium nitrate, saltpeter, nitre, nitrate of potash, vicknite, nitric acid potassium salt, nitric acid, potassium salt, saltpetre, kaliumnitrat, kalii nitras
CHEBI:63043
[N+](=O)([O-])[O-].[K+]
Safety and Handling

Safety and Handling

H272-H303: May intensify fire; oxidizer. May be harmful if swallowed.

DOTInformation : Hazard Class: 5.1, Packing Group: III

EINECSNumber : 231-818-8

Documents