missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium nitrate, 99.997% (metals basis), Alfa Aesar™ Puratronic™

Manufacturer:  Thermo Fisher Scientific 1084406

 View more versions of this product

Catalog No. AA1084406

Due to product restrictions, we cannot sell this product online.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Potassium nitrate
99.998% (metals basis)
MFCD00011405
asmofug e
[K+].[I-]
166.003
CHEBI:8346
Powder
1.33 hpa (1mm Hg) at 745°C
5g
7681-11-0
IK
White crystalline powder
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium;iodide
4875
166.01
Odorless
White

Chemical Identifiers

7681-11-0
166.003
NLKNQRATVPKPDG-UHFFFAOYSA-M
4875
potassium;iodide
IK
MFCD00011405
asmofug e
CHEBI:8346
[K+].[I-]
Documents