missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Nitrate, 10%, pH 12, Filling Solution for Cyanide ISE, Ricca Chemical

Chemical Identifiers

CAS 7732-18-5,7757-79-1,1310-58-3
CAS Min % 89.0,8.8,0.08
CAS Max % 93.0,9.16,0.08
Physical Form Liquid
Chemical Name or Material Potassium Nitrate, pH 12
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
R63190004A Ricca Chemical Company
RIR63190004A
Plastic Bottle 4L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7757-79-1,1310-58-3
93.0,9.16,0.08
Potassium Nitrate, pH 12
89.0,8.8,0.08
Liquid

Specifications

7732-18-5,7757-79-1,1310-58-3
93.0,9.16,0.08
Plastic Bottle
4L
89.0,8.8,0.08
Liquid
Clear colorless
Potassium Nitrate, pH 12