Please login to your online account to display your discounted pricing

Potassium Nitrate, 10%, pH 12, Filling Solution for Cyanide ISE, Ricca Chemical


Quantity 4L
Packaging Plastic Bottle

Manufacturer: ricca chemical company  RIR63190004A

 View more versions of this product

Catalog No. R63190004A

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Chemical Name or Material Potassium Nitrate
Name Note pH 12
CAS 7732-18-5
CAS Min % 89.0
CAS Max % 93.0
CAS 7757-79-1
CAS Min % 8.8
CAS Max % 9.16
CAS 1310-58-3
CAS Min % 0.08
CAS Max % 0.08
Physical Form Liquid
Packaging Plastic Bottle
Color Clear colorless
Quantity 4L

Specifications

Chemical Name or Material Potassium Nitrate
Name Note pH 12
CAS 7732-18-5
CAS Min % 89.0
CAS Max % 93.0
CAS 7757-79-1
CAS Min % 8.8
CAS Max % 9.16
CAS 1310-58-3
CAS Min % 0.08
CAS Max % 0.08
Physical Form Liquid