Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium niobium oxide, Puratronic™, 99.999% (metals basis)

$162.73 - $162.73

Chemical Identifiers

Chemical Name or Material Potassium niobium oxide
CAS 12030-85-2
Purity Grade Puratronic™
Formula Weight 180.01
MDL Number MFCD00049651
Synonym Potassium niobate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1084322

  SDS
alfa aesar™
1084322
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1084314

  SDS
alfa aesar™
1084314
25g Each for $162.73
Description & Specifications

Specifications

Formula Weight 180.01
Chemical Name or Material Potassium niobium oxide
TSCA TSCA
Purity Grade Puratronic™
Physical Form Powder
Assay 99.999%
Assay Percent Notes (metals basis)
MDL Number MFCD00049651
CAS 12030-85-2
Synonym Potassium niobate
Quantity 100g
RTECS Number TT3500000