Learn More
Potassium molybdenum oxide, anhydrous, 99.8% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 02289818
Description
Potassium molybdenum oxide used in chemical research, Chemicals, pharmaceutical research, pharmaceutical intermediates reagents, Laboratory chemicals, Manufacture of substances. And it is an important raw material and intermediates used in organic synthesis, agrochemicals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Potassium molybdenum oxide, anhydrous | |
13446-49-6 | |
Powder | |
99.8% (metals basis) | |
MFCD00011370 | |
Soluble in water. | |
[K+].[K+].[O-][Mo]([O-])(=O)=O | |
238.14 | |
238.14 | |
White powder |
919°C | |
White | |
50 g | |
K2MoO4 | |
potassium molybdenum oxide, dipotassium dioxido dioxo molybdenum, dipotassium ion molybdate, 2k.moo4, potassium molybdenum oxide, anhydrous | |
NUYGZETUIUXJCO-UHFFFAOYSA-N | |
dipotassium;dioxido(dioxo)molybdenum | |
6093816 | |
Odorless | |
2.3 g/cm3 |
Chemical Identifiers
13446-49-6 | |
238.14 | |
NUYGZETUIUXJCO-UHFFFAOYSA-N | |
6093816 |
K2MoO4 | |
MFCD00011370 | |
potassium molybdenum oxide, dipotassium dioxido dioxo molybdenum, dipotassium ion molybdate, 2k.moo4, potassium molybdenum oxide, anhydrous | |
[K+].[K+].[O-][Mo]([O-])(=O)=O |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 236-599-2
RTECSNumber : QA4930000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only