missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Methanesulfonate, TCI America™

Manufacturer:  TCI America M0550100G

 View more versions of this product

Catalog No. M0550100G



Specifications

Specifications

Potassium Methanesulfonate
100g
CH3KO3S
potassium methanesulfonate, methanesulfonic acid, potassium salt, potassium mesylate, potassium methylsulfonate, potassium methyl sulfonate, potassium methanesulphonate, unii-s99ib3em17, methanesulfonic acid potassium salt, methanesulfonic acid,potassium salt 1:1, methanesulfonic acid, potassium salt 1:1
CS(=O)(=O)[O-].[K+]
134.19
134.19
≥98.0% (T)
White
2386-56-3
MFCD00070544
XWIJIXWOZCRYEL-UHFFFAOYSA-M
potassium;methanesulfonate
23666501
Crystal-Powder at 20°C

Chemical Identifiers

2386-56-3
134.19
XWIJIXWOZCRYEL-UHFFFAOYSA-M
23666501
CS(=O)(=O)[O-].[K+]
CH3KO3S
MFCD00070544
potassium methanesulfonate, methanesulfonic acid, potassium salt, potassium mesylate, potassium methylsulfonate, potassium methyl sulfonate, potassium methanesulphonate, unii-s99ib3em17, methanesulfonic acid potassium salt, methanesulfonic acid,potassium salt 1:1, methanesulfonic acid, potassium salt 1:1
potassium;methanesulfonate
Safety and Handling

Safety and Handling

TSCA : Yes

Documents