Please login to your online account to display your discounted pricing

Potassium meta-Bisulfite, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500g
Packaging Poly Bottle

Manufacturer: avantor performance  297601

 View more versions of this product

Catalog No. 02003780

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Crystals
Formula Weight 222.33
Chemical Name or Material Potassium meta-Bisulfite
CAS 16731-55-8
Packaging Poly Bottle
Purity Grade BAKER ANALYZED™ Reagent
Specific Gravity 1L = 2.30kg
Molecular Formula K2S2O5
Quantity 500g

Specifications

Chemical Name or Material Potassium meta-Bisulfite
CAS 16731-55-8
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula K2S2O5
Formula Weight 222.33