Please login to your online account to display your discounted pricing

Potassium meta-Bisulfite, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$336.46 - $741.00

Chemical Identifiers

Chemical Name or Material Potassium meta-Bisulfite
CAS 16731-55-8
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula K2S2O5
Formula Weight 222.33
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003780

 
avantor performance
297601
Poly Bottle 500g Each for $336.46

02003781

 
avantor performance
297605
Poly Bottle 2.5kg Each for $741.00
Description & Specifications

Specifications

Physical Form Crystals
Formula Weight 222.33
Chemical Name or Material Potassium meta-Bisulfite
CAS 16731-55-8
Packaging Poly Bottle
Purity Grade BAKER ANALYZED™ Reagent
Specific Gravity 1L = 2.30kg
Molecular Formula K2S2O5
Quantity 500g