missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium (iodomethyl)trifluoroborate, tech. 90%

$78.50 - $299.18

Chemical Identifiers

CAS 888711-47-5
Molecular Formula CH2BF3IK
Molecular Weight (g/mol) 247.835
MDL Number MFCD11112187
InChI Key WQJVWRYIXQXISQ-UHFFFAOYSA-N
Synonym potassium trifluoro iodomethyl borate, potassium iodomethyltrifluoroborate, potassium trifluoro iodomethyl boranuide, pubchem11563, potassium iodomethyl trifluoroborate, trifluoro iodomethyl potassioboron v, potassium trifluoro iodomethyl borate 1-, potassium ion trifluoro iodomethyl boranuide, potassium tris fluoranyl-iodanylmethyl boranuide, borate 1-,trifluoro iodomethyl-, potassium, t-4-9ci
PubChem CID 23674071
IUPAC Name potassium;trifluoro(iodomethyl)boranuide
SMILES [B-](CI)(F)(F)F.[K+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6383903 View Documents Alfa Aesar™
H6383903
1g Each for $78.50
 
AAH6383906 View Documents Alfa Aesar™
H6383906
5g Each for $299.18
 
Description

Description

Applications
Organotrifluoroborate involved in synthesis of functionalized ethyltrifluoroborates, Suzuki-Miyaura cross-coupling reactions.

Notes
Light Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Specifications

Chemical Identifiers

888711-47-5
247.835
WQJVWRYIXQXISQ-UHFFFAOYSA-N
23674071
[B-](CI)(F)(F)F.[K+]
CH2BF3IK
MFCD11112187
potassium trifluoro iodomethyl borate, potassium iodomethyltrifluoroborate, potassium trifluoro iodomethyl boranuide, pubchem11563, potassium iodomethyl trifluoroborate, trifluoro iodomethyl potassioboron v, potassium trifluoro iodomethyl borate 1-, potassium ion trifluoro iodomethyl boranuide, potassium tris fluoranyl-iodanylmethyl boranuide, borate 1-,trifluoro iodomethyl-, potassium, t-4-9ci
potassium;trifluoro(iodomethyl)boranuide

Specifications

888711-47-5
ICH2BF3K
potassium trifluoro iodomethyl borate, potassium iodomethyltrifluoroborate, potassium trifluoro iodomethyl boranuide, pubchem11563, potassium iodomethyl trifluoroborate, trifluoro iodomethyl potassioboron v, potassium trifluoro iodomethyl borate 1-, potassium ion trifluoro iodomethyl boranuide, potassium tris fluoranyl-iodanylmethyl boranuide, borate 1-,trifluoro iodomethyl-, potassium, t-4-9ci
[B-](CI)(F)(F)F.[K+]
247.835
247.83
90%
Light Sensitive
242°C to 258°C
Potassium (iodomethyl)trifluoroborate
CH2BF3IK
MFCD11112187
WQJVWRYIXQXISQ-UHFFFAOYSA-N
potassium;trifluoro(iodomethyl)boranuide
23674071
Solid
Technical
Brown to White
1g
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning