Learn More
Potassium (iodomethyl)trifluoroborate, tech. 90%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6383906
Description
Organotrifluoroborate involved in synthesis of functionalized ethyltrifluoroborates, Suzuki-Miyaura cross-coupling reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsOrganotrifluoroborate involved in synthesis of functionalized ethyltrifluoroborates, Suzuki-Miyaura cross-coupling reactions.
Notes
Light Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Specifications
Potassium (iodomethyl)trifluoroborate | |
Brown to White | |
Solid | |
CH2BF3IK | |
MFCD11112187 | |
WQJVWRYIXQXISQ-UHFFFAOYSA-N | |
potassium trifluoro(iodomethyl)boranuide | |
23674071 | |
90% | |
Light sensitive |
242°C to 258°C | |
888711-47-5 | |
5 g | |
ICH2BF3K | |
potassium trifluoro iodomethyl borate, potassium iodomethyltrifluoroborate, potassium trifluoro iodomethyl boranuide, pubchem11563, potassium iodomethyl trifluoroborate, trifluoro iodomethyl potassioboron v, potassium trifluoro iodomethyl borate 1-, potassium ion trifluoro iodomethyl boranuide, potassium tris fluoranyl-iodanylmethyl boranuide, borate 1-,trifluoro iodomethyl-, potassium, t-4-9ci | |
[K+].F[B-](F)(F)CI | |
247.83 | |
247.83 | |
Technical |
Chemical Identifiers
888711-47-5 | |
247.83 | |
WQJVWRYIXQXISQ-UHFFFAOYSA-N | |
23674071 |
CH2BF3IK | |
MFCD11112187 | |
potassium trifluoro iodomethyl borate, potassium iodomethyltrifluoroborate, potassium trifluoro iodomethyl boranuide, pubchem11563, potassium iodomethyl trifluoroborate, trifluoro iodomethyl potassioboron v, potassium trifluoro iodomethyl borate 1-, potassium ion trifluoro iodomethyl boranuide, potassium tris fluoranyl-iodanylmethyl boranuide, borate 1-,trifluoro iodomethyl-, potassium, t-4-9ci | |
[K+].F[B-](F)(F)CI |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only