missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium iodide, ultra dry, 99.998% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 1367614

 View more versions of this product

Catalog No. AA1367614



Specifications

Specifications

Potassium iodide
99.998% (metals basis)
MFCD00011405
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
[K+].[I-]
166.003
CHEBI:8346
Beads
1.33 hPa (1mm Hg) at 745°C
25g
7681-11-0
IK
White beads
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium;iodide
4875
166.01
Odorless
White

Chemical Identifiers

7681-11-0
166.003
NLKNQRATVPKPDG-UHFFFAOYSA-M
4875
potassium;iodide
IK
MFCD00011405
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
CHEBI:8346
[K+].[I-]
Safety and Handling

Safety and Handling

EINECSNumber : 213-659-4

Documents