Please login to your online account to display your discounted pricing

Potassium iodide, ultra dry, 99.998% (metals basis), Alfa Aesar™

$64.93 - $64.93

Chemical Identifiers

Chemical Name or Material Potassium iodide
CAS 7681-11-0
Physical Form Beads
Assay Percent Range 99.998% (metals basis)
Molecular Formula KI
Formula Weight 166.01
MDL Number MFCD00011405
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1367606

  SDS
alfa aesar
1367606
5g Each for $64.93

AA1367614

  SDS
alfa aesar
1367614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Beads
Formula Weight 166.01
Quantity 5g
Color White
Density 3.12g/cm3
Chemical Name or Material Potassium iodide
EINECS Number 213-659-4
Odor Odorless
Assay Percent Range 99.998% (metals basis)
Molecular Formula KI
Appearance White beads
MDL Number MFCD00011405
CAS 7681-11-0
Vapor Pressure 1.33 hPa (1mm Hg) at 745°C