missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Iodide Solution (10% W/v), Fisher Chemical

Manufacturer:  Fisher Scientific Company FLSP242-500

 View more versions of this product

Catalog No. SP242-500



 

Specifications Description & Specifications

Specifications

Potassium Iodide Solution
7681-11-0,7732-18-5
9 to 11%
1.032g/cm³
10% w/v
Glass Bottle
Clear
10% (w/v)
Liquid
10% w/v
MFCD00011405
Pass Test
14mmHg
500mL

Chemical Identifiers

7681-11-0,7732-18-5
MFCD00011405
9 to 11%
10% w/v
Potassium Iodide Solution, 10% (w/v)

Safety and Handling

SDS & Safety Documents

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Obtain medical attention. Obtain medical attention. .

NFPA

Health:1
Flammability:0
Instability:0