Please login to your online account to display your discounted pricing

Potassium Iodide-Iodine Reagent, LabChem™

$105.60 - $105.60

Chemical Identifiers

Chemical Name or Material Potassium Iodide-Iodine Reagent
CAS 7732-18-5
CAS Max % 98.75
CAS 7681-11-0
CAS Max % 0.75
CAS 7553-56-2
CAS Max % 0.5
Linear Formula I2
Formula Weight 253.809
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC198162

 
lab chem
LC198162
1L Each for $105.60
Description & Specifications

Specifications

Formula Weight 253.809
Linear Formula I2
Color Dark red-brown
Health Hazard 2 GHS H Statement
Harmful to aquatic life.
Packaging Amber Glass
CAS 7732-18-5
CAS 7681-11-0
Quantity 1L
CAS Max % 0.5
CAS Max % 0.75
Identification Passes Test
Storage Information Room Temperature
CAS 7553-56-2
CAS Max % 98.75
Health Hazard 3 GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.
Chemical Name or Material Potassium Iodide-Iodine Reagent
Solubility Soluble in water
Decomposition Information Iodine vapor; Potassium oxide
Physical Form Liquid
Safety and Handling

GHS P Statement
Avoid release to the environment.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful to aquatic life.