missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium Iodide (Granular, Free Flowing/USP/FCC), Fisher Chemical™
Supplier: Fisher Chemical FLP41210
Specifications
1330°C | |
680°C | |
White | |
Solid | |
IK | |
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
[K+].[I-] | |
166.003 | |
CHEBI:8346 | |
99.0 to 101.5% (FCC) | |
Pass Test (USP/FCC), Pass Test (FCC), Pass Test (USP) | |
Poly Pail |
Potassium Iodide | |
7681-11-0 | |
6 to 8 | |
10 kg | |
MFCD00011405 | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
potassium;iodide | |
4875 | |
166 | |
FCC/USP | |
1% max. (FCC), 1.0% max. (USP), 1.0% max. (USP), 1% max. (FCC) | |
1mmHg at 745°C |
Chemical Identifiers
7681-11-0 | |
166.003 | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
4875 | |
potassium;iodide |
IK | |
MFCD00011405 | |
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
CHEBI:8346 | |
[K+].[I-] |
Safety and Handling
CAUTION!
Emergency Overview
May cause skin, eye, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:1
Flammability:1
Instability:0