Please login to your online account to display your discounted pricing

Potassium Iodide, For Arsenic, Certified, 15.0% (w/v) ±0.5% (w/v), LabChem™

$68.11 - $1,027.09

Chemical Identifiers

Chemical Name or Material Potassium Iodide
CAS 7732-18-5
CAS Max % 85
CAS 7681-11-0
CAS Max % 15
Name Note For Arsenic
Purity Grade Certified Grade
Linear Formula KI
Formula Weight 166
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC197801

 
lab chem
LC197801
500mL Each for $68.11

LC197802

 
lab chem
LC197802
1L Each for $107.63

LC197804

 
lab chem
LC197804
4L Each for $334.60

LC197805

 
lab chem
LC197805
20L Each for $1,027.09

LC197806

 
lab chem
LC197806
10L Each for $765.54
Description & Specifications

Specifications

Purity Grade Certified Grade
Assay 15.0% ± 0.5% (w/v)
Formula Weight 166
Chemical Name or Material Potassium Iodide
Traceability to NIST Traceable to NIST
Viscosity 0.84cSt
Decomposition Information Iodine vapor
Color Clear to slightly yellow
Quantity 500mL
Name Note For Arsenic
Physical Form Liquid
Linear Formula KI
Density 1.11g/mL
CAS 7732-18-5
Health Hazard 1 Warning
CAS Max % 85
Health Hazard 2 GHS H Statement
Causes eye irritation.
Identification Passes Test
CAS 7681-11-0
Health Hazard 3 GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Packaging Amber Poly
CAS Max % 15
Solubility Soluble in water
Specific Gravity 1.11g/mL
Storage Information Room Temperature; Protect from light
Safety and Handling

Warning

GHS P Statement
Wash exposed skin thoroughly after handling.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
Causes eye irritation.