Please login to your online account to display your discounted pricing

Potassium Iodide, Certified, 2.0% (w/v) ±0.1%, LabChem™

$59.03 - $59.03

Chemical Identifiers

Chemical Name or Material Potassium Iodide
CAS 7732-18-5
CAS Max % 98
CAS 7681-11-0
CAS Max % 2
Purity Grade Certified Grade
Linear Formula KI
Formula Weight 166
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC197201

 
lab chem
LC197201
500mL Each for $59.03
Description & Specifications

Specifications

Assay 2.0% ± 0.1% (w/v)
Purity Grade Certified Grade
Formula Weight 166
Solubility Soluble in water
CAS Max % 2
Storage Information Room Temperature; Protect from light
Quantity 500mL
Decomposition Information Iodine vapor
Viscosity 0.98cSt
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Linear Formula KI
Packaging Amber Poly
CAS 7732-18-5
Physical Form Liquid
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Specific Gravity 1.01g/mL
Identification Passes Test
Color Clear, colorless
CAS 7681-11-0
CAS Max % 98
Chemical Name or Material Potassium Iodide
Density 1.01g/mL
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.