missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Iodide, ACS Reagent Grade, Ricca Chemical

$763.03 - $763.03

Chemical Identifiers

CAS 7681-11-0
CAS Min % 98.0
CAS Max % 100.0
Physical Form Powder or Crystals
Chemical Name or Material Potassium Iodide
Grade ACS Reagent Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
RDCP05603A5 Ricca Chemical Company
RIRDCP05603A5
Poly natural 3kg Each for $763.03
RDCP0560100 Ricca Chemical Company
RIRDCP0560100B1
Poly Wide mouth 100 gal. N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
RDCP0560500 Ricca Chemical Company
RIRDCP0560500B1
Poly Wide mouth 500 gal. N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7681-11-0
100.0
Potassium Iodide
98.0
Powder or Crystals
ACS Reagent Grade

Specifications

7681-11-0
100.0
Poly natural
3kg
ACS Reagent Grade
98.0
Powder or Crystals
White
Potassium Iodide