Please login to your online account to display your discounted pricing

Potassium Iodide ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Potassium Iodide
Name Note Free Flowing
CAS 7681-11-0
Purity Grade ACS Grade
Molecular Formula KI
Formula Weight 166
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK1123500

 
avantor performance
112312
POLYSTORMOR 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK1123212

 
avantor performance
112306
Poly 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 166
Chemical Name or Material Potassium Iodide
Name Note Free Flowing
Packaging POLYSTORMOR
Molecular Formula KI
CAS 7681-11-0
Quantity 500g