missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Iodide ACS AR Granular, Macron Fine Chemicals™

macron

Chemical Identifiers

CAS 7681-11-0
Physical Form Granular
Molecular Formula KI
Formula Weight 166
Chemical Name or Material Potassium Iodide, Neutral
Grade ACS Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
MK1127212 Avantor Performance
112706
Poly 2.5kg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7681-11-0
KI
Potassium Iodide, Neutral
Granular
166
ACS Grade

Specifications

7681-11-0
KI
Poly
Potassium Iodide, Neutral
Granular
166
2.5kg
ACS Grade