Please login to your online account to display your discounted pricing

Potassium iodide, ACS, 99.0% min, Alfa Aesar™

In photographic emulsions, in analytical chemistry, as a feed additive, in spectroscopy, in infrared transmission, and in scintillation

$715.77 - $715.77

Chemical Identifiers

Chemical Name or Material Potassium iodide
CAS 7681-11-0
Purity Grade ACS Grade
Molecular Formula KI
Formula Weight 166.01
MDL Number MFCD00011405
Merck Index 14,7643
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA11601A3

 
alfa aesar
11601A3
Each for $715.77
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 166.01
Quantity 2kg
Chemical Name or Material Potassium iodide
TSCA TSCA
Odor Odorless
Physical Form Granular
Refractive Index 1.677
Assay ≥99.0%
Sensitivity Hygroscopic
Molecular Formula KI
MDL Number MFCD00011405
CAS 7681-11-0
Merck Index 14,7643
Boiling Point 1330°C
Melting Point 681°C
Solubility Information Soluble in water, alcohol, acetone, and glycerol
Density 3.12
RTECS Number TT2975000