missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium iodide, 99%, extra pure, briquettes, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 196731000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
1330.0°C | |
680.0°C | |
White | |
100 g | |
IK | |
06,227; 11,44 | |
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
potassium;iodide | |
4875 | |
166 | |
Extra Pure | |
Glass bottle |
Potassium iodide | |
7681-11-0 | |
Briquettes | |
98.5% min. (on dried substance) (Argentometry) | |
KI | |
15,7764 | |
Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol | |
[K+].[I-] | |
166 | |
CHEBI:8346 | |
99% | |
1% max. |
Chemical Identifiers
7681-11-0 | |
166 | |
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
CHEBI:8346 | |
[K+].[I-] |
IK | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
4875 | |
potassium;iodide |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 231-659-4
RUO – Research Use Only