missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Potassium iodide, 99.995%, (trace metal basis), extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 193790100
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisSpecifications
1330.0°C | |
680.0°C | |
White | |
10 g | |
IK | |
MFCD00011405 | |
15, 7764 | |
Solubility in water: 1.43 kg/l. Other solubilities: 45g/L ethanol | |
[K+].[I-] | |
166 | |
CHEBI:8346 | |
99.995% | |
Glass bottle |
Potassium iodide | |
7681-11-0 | |
Crystals or Powder | |
99.995% | |
KI | |
06,227; 11,440 | |
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
potassium;iodide | |
4875 | |
166 | |
Trace Metal Basis |
Chemical Identifiers
7681-11-0 | |
166 | |
NLKNQRATVPKPDG-UHFFFAOYSA-M | |
4875 | |
potassium;iodide |
IK | |
MFCD00011405 | |
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide | |
CHEBI:8346 | |
[K+].[I-] |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 231-659-4
RUO – Research Use Only