Please login to your online account to display your discounted pricing

Potassium iodide, 99.9% (metals basis), Alfa Aesar™

$37.05 - $320.95

Chemical Identifiers

Chemical Name or Material Potassium iodide
CAS 7681-11-0
Physical Form Crystalline Powder
Assay Percent Range 99.9% (metals basis)
Molecular Formula KI
Formula Weight 166.01
MDL Number MFCD00011405
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4285714

  SDS
alfa aesar
4285714
25g Each for $37.05

AA4285736

  SDS
alfa aesar
4285736
500g Each for $320.95

AA4285722

  SDS
alfa aesar
4285722
100g Each for $89.51
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 166.01
Quantity 25g
Color White
Density 3.12g/cm3 at 20°C
EINECS Number 213-659-4
Chemical Name or Material Potassium iodide
CAS 7681-11-0
Vapor Pressure 1.33 hPa (1mm Hg) at 20°C
Odor Odorless
Assay Percent Range 99.9% (metals basis)
MDL Number MFCD00011405
Appearance White crystalline powder
Molecular Formula KI