missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium iodide, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A127040B

 View more versions of this product

Catalog No. AAA127040B



Description

Description

Potassium iodide is used in pharmaceutical formulations and in film photography as a source of iodine. It is widely used as an analytical Reagent in iodometric tritrations. Apart from its application as a source of iodide to produce organo iodine compounds such as in Sandmeyer reactions, potassium iodide finds extensive use as a nucleophilic catalyst for several alkylation reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Potassium iodide is used in pharmaceutical formulations and in film photography as a source of iodine. It is widely used as an analytical Reagent in iodometric tritrations. Apart from its application as a source of iodide to produce organo iodine compounds such as in Sandmeyer reactions, potassium iodide finds extensive use as a nucleophilic catalyst for several alkylation reactions.

Solubility
Soluble in water

Notes
Being a mild reducing agent, it gets converted into iodine by oxidizing reagents. Potassium iodide should be handled with gloves since it is a mild irritant.
Specifications

Specifications

1330°C
681°C
1000 g
MFCD00011405
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
NLKNQRATVPKPDG-UHFFFAOYSA-M
potassium;iodide
4875
166.01
Odorless
Air and light sensitive
Potassium iodide
7681-11-0
IK
14,7643
Soluble in water
[K+].[I-]
166.003
CHEBI:8346
99%
1.677
3.12

Chemical Identifiers

7681-11-0
166.003
NLKNQRATVPKPDG-UHFFFAOYSA-M
4875
potassium;iodide
IK
MFCD00011405
potassium iodide, potassium iodide ki, pima, kali iodide, thyro-block, knollide, kisol, asmofug e, potassium diiodide, potassium monoiodide
CHEBI:8346
[K+].[I-]
Safety and Handling

Safety and Handling

P260-P264b-P270-P314-P501c

H372

EINECSNumber : 231-659-4

RTECSNumber : TT2975000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only