missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Potassium Iodide, 5% (w/v) Aqueous Solution, Stabilized, Ricca Chemical

Chemical Identifiers

CAS 7732-18-5,7681-11-0,144-55-8
CAS Min % 93.0,4.67,0.01
CAS Max % 97.0,4.86,0.01
Physical Form Liquid
Chemical Name or Material Potassium Iodide
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
62902-5 Ricca Chemical Company
RI62902.5
Cubitainer 10L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
62905 SDS Ricca Chemical Company
RI62905
Cubitainer 20L N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7681-11-0,144-55-8
97.0,4.86,0.01
Potassium Iodide
93.0,4.67,0.01
Liquid

Specifications

7732-18-5,7681-11-0,144-55-8
97.0,4.86,0.01
Cubitainer
5% (w/v)
Clear colorless
Potassium Iodide
93.0,4.67,0.01
Liquid
Stabilized
Aqueous Solution
10L