Please login to your online account to display your discounted pricing

Potassium Iodide, 20% (w/v) Aqueous Solution, Stabilized, Ricca Chemical

$189.91 - $343.73

Chemical Identifiers

Chemical Name or Material Potassium Iodide
CAS 7681-11-0 (Potassium Iodide)
CAS 7732-18-5 (Water, Deionized)
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

6310-32

  SDS
ricca chemical company
RI631032
Amber Poly 1L Each for $343.73

6310-16

  SDS
ricca chemical company
RI631016
Amber Poly 500mL Each for $189.91

6310-5

  SDS
ricca chemical company
RI63105
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless to slightly yellow
Physical Form Liquid
Quantity 1L
Treatment(s) Stabilized
Solvent or Matrix Aqueous Solution
Concentration or Composition Notes 20% (w/v)
Chemical Name or Material Potassium Iodide
CAS 7681-11-0 (Potassium Iodide)
CAS 7732-18-5 (Water, Deionized)
Packaging Amber Poly