Please login to your online account to display your discounted pricing

Potassium Iodide, 2% (w/v) Aqueous Solution, Stabilized, APHA for Ozone, Ricca Chemical

$72.00 - $72.00

Chemical Identifiers

Chemical Name or Material Potassium Iodide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

6288-16

 
ricca chemical company
RI628816
Amber Plastic 500mL Each for $72.00

6288-32

 
ricca chemical company
RI628832
Amber Plastic 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 500mL
Methods APHA for Ozone
Treatment(s) Stabilized
Chemical Name or Material Potassium Iodide
Packaging Amber Plastic
Concentration or Composition (by Analyte or Components) 2% (w/v) Aqueous Solution