Please login to your online account to display your discounted pricing

Potassium Iodide, 2% (w/v) Aqueous Solution, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Potassium Iodide
CAS 7732-18-5
CAS Min % 96.0
CAS Max % 100.0
CAS 7681-11-0
CAS Min % 1.92
CAS Max % 2.0
CAS 144-55-8
CAS Min % 0.01
CAS Max % 0.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

62885

 
ricca chemical company
RI62885
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R62880004B

 
ricca chemical company
RIR62880004B
Amber poly 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 20L
Chemical Name or Material Potassium Iodide
Concentration or Composition Notes 2% (w/v)
Solvent or Matrix Aqueous Solution
CAS 7732-18-5
CAS Max % 100.0
CAS Max % 0.01
CAS Min % 96.0
CAS 7681-11-0
Packaging Cubitainer
CAS Min % 1.92
CAS Max % 2.0
CAS 144-55-8
CAS Min % 0.01