Please login to your online account to display your discounted pricing

Potassium Iodide, 16% (w/v) Aqueous Solution, Ricca Chemical

$1,386.65 - $1,386.65

Chemical Identifiers

Chemical Name or Material Potassium Iodide
CAS 7732-18-5
CAS Min % 84.0
CAS Max % 88.0
CAS 7681-11-0
CAS Min % 13.52
CAS Max % 14.08
CAS 144-55-8
CAS Min % 0.01
CAS Max % 0.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

6306-5

 
ricca chemical company
RI63065
Cubitainer 20L Each for $1,386.65

6306-1

  SDS
ricca chemical company
RI63061
Amber Poly 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless to slightly yellow
Quantity 20L
Concentration or Composition Notes 16% (w/v)
Solvent or Matrix Aqueous Solution
CAS Min % 84.0
CAS Max % 88.0
Chemical Name or Material Potassium Iodide
CAS Max % 0.01
CAS Min % 0.01
CAS 144-55-8
CAS Min % 13.52
CAS Max % 14.08
Packaging Cubitainer
CAS 7681-11-0
CAS 7732-18-5