Please login to your online account to display your discounted pricing

Potassium Iodide, 15% (w/v) Aqueous Solution, Stabilized, Ricca Chemical

$143.94 - $682.76

Chemical Identifiers

Chemical Name or Material Potassium Iodide
CAS 7732-18-5
CAS Min % 85.0
CAS Max % 89.0
CAS 7681-11-0
CAS Min % 12.79
CAS Max % 13.31
CAS 144-55-8
CAS Min % 0.01
CAS Max % 0.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

6305-1

 
ricca chemical company
RI63051
Amber Poly 4L Each for $682.76

6305-16

 
ricca chemical company
RI630516
Amber Poly 500mL Each for $143.94

6305-32

 
ricca chemical company
RI630532
Amber Poly 1L Each for $232.09

63052-5

  SDS
ricca chemical company
RI63052.5
Cubitainer 10L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

63055

  SDS
ricca chemical company
RI63055
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless to slightly yellow
Quantity 4L
Solvent or Matrix Aqueous Solution
Concentration or Composition Notes 15% (w/v)
Treatment(s) Stabilized
CAS Min % 85.0
CAS Max % 89.0
Chemical Name or Material Potassium Iodide
Packaging Amber Poly
CAS Min % 12.79
CAS 144-55-8
CAS Max % 13.31
CAS Max % 0.01
CAS Min % 0.01
CAS 7732-18-5
CAS 7681-11-0