Please login to your online account to display your discounted pricing

Alfa Aesar™ Potassium iodide, 1:2 w/w soln. in distilled water

Chemical Identifiers

Chemical Name or Material Potassium iodide
CAS 7681-11-0
Formula Weight 166.01
MDL Number MFCD00011405
Merck Index 14,7643
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA47293K2

 
alfa aesar™
47293K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 166.01
Chemical Name or Material Potassium iodide
TSCA TSCA
Physical Form Liquid
MDL Number MFCD00011405
CAS 7681-11-0
Merck Index 14,7643
Odor Odorless
Refractive Index 1.677
Density 3.12g/mL
Boiling Point 1330°C
Color Colorless
Melting Point 681°C
Concentration 1:2 w/w soln. in distilled water
Quantity 1L
Sensitivity Hygroscopic
RTECS Number TT2975000