Please login to your online account to display your discounted pricing

Potassium Iodide, 1.0N, Fisher Chemical


Quantity 500mL
Packaging Glass Bottle

Manufacturer: fisher scientific company  SP821500

 View more versions of this product

Catalog No. SP821-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Quantity 500mL
Viscosity [NULL]
Vapor Pressure 14mmHg
NIST Traceable to NIST.
Color Clear
Physical Form Liquid
Density 1.032g/cm³
MDL Number MFCD00011405
ChemAlert Storage Symbol Gray
Packaging Glass Bottle
Chemical Name or Material Potassium Iodide
Name Note 1.0N
Concentration 0.995 to 1.005N
CAS 7732-18-5
Concentration or Composition (by Analyte or Components) 1.0N
CAS 7681-11-0

Specifications

Chemical Name or Material Potassium Iodide
Name Note 1.0N
CAS 7732-18-5
CAS 7681-11-0
MDL Number MFCD00011405
Accessories
 

SDS & Safety Documents