Please login to your online account to display your discounted pricing

Potassium Iodide, 1.00 Normal, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Potassium Iodide
CAS 7732-18-5
CAS Min % 84.0
CAS Max % 87.0
CAS 7681-11-0
CAS Min % 13.96
CAS Max % 14.53
CAS 144-55-8
CAS Min % 0.01
CAS Max % 0.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

6318-16

 
ricca chemical company
RI631816
Glass amber 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

63185

  SDS
ricca chemical company
RI63185
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless to slightly yellow
Quantity 500mL
Chemical Name or Material Potassium Iodide
CAS 7732-18-5
Concentration or Composition Notes 1.00Normal
CAS 7681-11-0
CAS Min % 13.96
CAS Max % 0.01
CAS Min % 0.01
CAS Max % 87.0
CAS Min % 84.0
CAS 144-55-8
CAS Max % 14.53
Packaging Glass amber