Please login to your online account to display your discounted pricing

Potassium Iodide, 0.5% (w/v) Aqueous Solution, Stabilized, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Potassium Iodide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

6287-16

 
ricca chemical company
RI628716
Amber Plastic 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

6287-4

 
ricca chemical company
RI62874
Amber Plastic 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 500mL
Treatment(s) Stabilized
Chemical Name or Material Potassium Iodide
Concentration or Composition (by Analyte or Components) 0.5% (w/v) Aqueous Solution
Packaging Amber Plastic